Merck
Inhibitory effect of asiatic acid on acetylcholinesterase, excitatory post synaptic potential and locomotor activity.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2011-11-25)
M N Nasir, J Abdullah, M Habsah, R I Ghani, G Rammes
RESUMEN
The asiatic acid, a triterpenoids isolated from Centella asiatica was used to delineate its inhibitory effect on acetylcholinesterase (AChE) properties, excitatory post synaptic potential (EPSP) and locomotor activity. This study is consistent with asiatic acid having an effect on AChE, a selective GABA(B) receptor agonist and no sedative effect on locomotor.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Asiatic acid, 97%
Sigma-Aldrich
Asiatic acid, ≥98% (HPLC), from Centella asiatica