Merck
  • Poly(4-vinylpyridinium)hydrogen sulfate: a novel and efficient catalyst for the synthesis of 14-aryl-14H-dibenzo[a,j]xanthenes under conventional heating and ultrasound irradiation.
Poly(4-vinylpyridinium)hydrogen sulfate: a novel and efficient catalyst for the synthesis of 14-aryl-14H-dibenzo[a,j]xanthenes under conventional heating and ultrasound irradiation.
Ultrasonics sonochemistry (2012-01-17)
Nader Ghaffari Khaligh
RESUMEN
A simple and convenient procedure for the synthesis of 14-aryl-14H-dibenzo[a,j]xanthenes is described through a one-pot condensation of 2-naphthol with aryl aldehydes in the presence of poly(4-vinylpyridinium)hydrogen sulfate as an efficient, cheap, readily synthesized and eco-friendly catalyst in a solvent free media using conventional heating and ultrasound irradiation.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Poly(4-vinylpyridine), average Mw ~60,000
Sigma-Aldrich
Poly(4-vinylpyridine), average Mw ~160,000