Merck
Safety and pharmacokinetics of nelfinavir during the second and third trimesters of pregnancy and postpartum.
HIV clinical trials (2012-02-07)
A Fang, S R Valluri, M-J O'Sullivan, R Maupin, T Jones, I Delke, P Clax
RESUMEN
Evaluate the safety, tolerability, and pharmacokinetics (PK) of nelfinavir during pregnancy and postpartum in HIV-infected women. Phase IV, non-randomized, open-label study of nelfinavir 625 mg tablets (1250 mg) in combination with lamivudine/zidovudine twice daily. Primary endpoint was treatment-related or possibly treatment-related gastrointestinal or hepatic adverse events (AEs). Selected maternal and infant outcomes were recorded. Frequent plasma samples were collected for PK studies during the 2nd and 3rd trimesters, and 6 weeks postpartum, to analyze total and free nelfinavir and M8 concentrations. Sixteen HIV+ pregnant women were enrolled. Six mild treatment-related AEs and 3 serious AEs occurred; 1 serious AE (elevated AST) met the primary endpoint. Compared with 6 weeks postpartum, levels of total nelfinavir were reduced by 44% and 46%, total M8 by 82% and 83%, free nelfinavir by 48% and 39%, and free M8 by 83% and 79% in the 2nd and 3rd trimesters, respectively. At 6 weeks postpartum, 75% and 50% of subjects maintained HIV-1 RNA levels <400 and <50 copies/mL, respectively. All pregnancies resulted in live births without transmission in 15 infants. Nelfinavir in combination with lamivudine/zidovudine was generally well tolerated. Total and free nelfinavir and M8 exposure were reduced in late pregnancy.