Merck
  • Efficacy of a commercial dentifrice containing 2% strontium chloride and 5% potassium nitrate for dentin hypersensitivity: a 3-day clinical study in adults in China.
Efficacy of a commercial dentifrice containing 2% strontium chloride and 5% potassium nitrate for dentin hypersensitivity: a 3-day clinical study in adults in China.
Clinical therapeutics (2012-03-06)
Hongchun Liu, Deyu Hu
RESUMEN
Studies of dentifrices containing strontium chloride or potassium nitrate have documented the clinical efficacy of these formulations for dentin hypersensitivity (DH), but few studies have evaluated dentifrices containing both active ingredients. The objective of this study was to compare the effects on DH of a dentifrice containing 2% strontium chloride and 5% potassium nitrate in a silica base (experimental dentifrice) with those of a dentifrice containing the same silica base without any active ingredient (control dentifrice). Male and female patients aged 20 to 65 years with DH were eligible to participate in this randomized, double-blind, placebo-controlled study in China. Patients were assigned to receive the experimental or control dentifrice. At baseline, immediately after topical dentifrice use, and after 3 days, patients received an oral examination of their hard and soft tissues, followed by an evaluation of DH that used the Yeaple Probe to measure tactile hypersensitivity and the Schiff Cold Air Scale to measure the perception of pain from an air blast stimulus. Adverse events (in particular, discomfort while brushing or alteration in taste) in the oral hard or soft tissues were monitored throughout the study. A total of 81 patients were enrolled (40 in the experimental group; 41 in the control group), of whom 79 (55 women; 24 men) completed the study. After topical use, the experimental group had a significantly higher mean (SD) tactile hypersensitivity score (19.47 [14.69] vs 14.27 [5.76]; P = 0.047) and a significantly lower mean air blast hypersensitivity score (1.93 [0.51] vs 2.22 [0.60]; P = 0.026) than did the control group. After 3 days, the experimental group had a significantly higher mean tactile hypersensitivity score (19.87 [14.95] vs 14.51 [6.00]; P = 0.045) and a significantly lower air blast hypersensitivity score (1.80 [0.56] and 2.13 [0.60]; P = 0.014) than the control group. After topical use and after 3 days, the experimental group had increases from baseline in tactile scores of 54.04% and 56.67%, respectively (both, P = 0.001) and reductions from baseline in air blast scores of 18.51% and 24.21% (both, P < 0.001); the control group had increases in tactile scores of 21.14% and 21.54% (P = 0.022 and P = 0.007) and reductions in air blast scores of 10.24% and 13.41% (P = 0.001 and P < 0.001). No adverse events were reported throughout the study. In these patients with DH in China, the dentifrice containing 2% strontium chloride and 5% potassium nitrate was efficacious in reducing DH when used instantly after topical dentifrice use and after 3 days of use.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Potassium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Strontium chloride hexahydrate, ACS reagent, 99%
Sigma-Aldrich
Potassium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Potassium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Strontium chloride, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Strontium chloride hexahydrate, 99.995% trace metals basis
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Supelco
Mettler-Toledo Calibration substance ME 51143095, Potassium nitrate, traceable to primary standards (LGC)
Sigma-Aldrich
Strontium chloride solution, 0.1 M