Merck
  • The composition of the culture medium influences the β-1,3-glucan metabolism of Aspergillus fumigatus and the antifungal activity of inhibitors of β-1,3-glucan synthesis.
The composition of the culture medium influences the β-1,3-glucan metabolism of Aspergillus fumigatus and the antifungal activity of inhibitors of β-1,3-glucan synthesis.
Antimicrobial agents and chemotherapy (2012-03-07)
Cécile Clavaud, Anne Beauvais, Lise Barbin, Hélène Munier-Lehmann, Jean-Paul Latgé
RESUMEN
In vitro testing of Aspergillus fumigatus susceptibility to echinocandins has always been a challenge. Using a simple and quick colorimetric method to analyze the activity of inhibitors of β-1,3-glucan synthesis, we found that the composition of the culture medium significantly influences glucan synthesis and consequently the antifungal properties of inhibitors of β-1,3-glucan synthesis when they are tested alone or in combination with chitin synthase inhibitors.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
β-1,3-Glucan from Euglena gracilis