Merck
  • Ultraviolet Resonance Raman spectroscopy used to study formulations of salmon calcitonin, a starch-peptide conjugate and TGF-β3.
Ultraviolet Resonance Raman spectroscopy used to study formulations of salmon calcitonin, a starch-peptide conjugate and TGF-β3.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2012-03-17)
E Patois, I A Larmour, S E J Bell, C Palais, M A H Capelle, R Gurny, T Arvinte
RESUMEN
Ultraviolet Resonance Raman (UVRR) spectroscopy with excitation at 244 nm was investigated here as a possible useful tool for fast characterization of biopharmaceuticals. Studies were performed on three protein drugs: salmon calcitonin (sCT), starch-peptide conjugate, and transforming growth factor-β3 (TGF-β3) adsorbed onto solid granules of tricalcium phosphate (TCP). Secondary structure of sCT was investigated for solutions of 0.5mg/mL up to 200mg/mL, regardless of the turbidity or aggregation states. An increase in β-sheet content was detected when sCT solutions aggregated. UVRR spectroscopy also detected a small amount of residual organic solvent in a starch-peptide conjugate solution containing only 40 μg/mL of peptide. UVRR spectroscopy was then used to characterize a protein, TGF-β3, adsorbed onto solid granules of TCP at 50 and 250 μg/cm(3). This study shows that UVRR is suitable to characterize the protein formulations in a broad range of concentrations, in liquid, aggregated, and solid states.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Calcitonin salmon, ≥97% (HPLC), powder
Calcitonin salmon, European Pharmacopoeia (EP) Reference Standard