Merck
  • Comparison of nitroethane, 2-nitro-1-propanol, lauric acid, Lauricidin® and the Hawaiian marine algae, Chaetoceros, for potential broad-spectrum control of anaerobically grown lactic acid bacteria.
Comparison of nitroethane, 2-nitro-1-propanol, lauric acid, Lauricidin® and the Hawaiian marine algae, Chaetoceros, for potential broad-spectrum control of anaerobically grown lactic acid bacteria.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2012-03-21)
Aleksandar K Božic, Robin C Anderson, Steven C Ricke, Philip G Crandall, Corliss A O'Bryan
RESUMEN
The gastrointestinal tract of bovines often contains bacteria that contribute to disorders of the rumen, and may also contain foodborne or opportunistic human pathogens as well as bacteria capable of causing mastitis in cows. Thus there is a need to develop broad-spectrum therapies that are effective while not leading to unacceptably long antibiotic withdrawal times. The effects of the CH(4)-inhibitors nitroethane (2 mg/mL), 2-nitro-1-propanol (2 mg/mL), lauric acid (5 mg/mL), the commercial product Lauricidin® (5 mg/mL), and a finely ground product of the Hawaiian marine algae, Chaetoceros (10 mg/mL), were compared in pure cultures of Streptococcus agalactia, Enterococcus faecium, Streptococcus bovis, and in a mixed lactic acid rumen bacterial culture. Lauricidin® and lauric acid exhibited the most bactericidal acidity against all bacteria. These results suggest potential animal health benefits from supplementing cattle diets with lauric acid or Lauricidin® to improve the health of the rumen and help prevent shedding of human pathogens.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Dodecanoic acid, 98%
Sigma-Aldrich
Sodium dodecanoate, 99-100%
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)
Supelco
Dodecanoic acid, analytical standard