Merck
Spectroscopic investigation on the interaction of 9-aminoacridine with certain dyes.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-05-16)
C Manivannan, R Renganathan
RESUMEN
The fluorescence quenching of 9-Aminoacridine (9-AA) by various dyes were probed in aqueous medium using steady state and time resolved fluorescence spectroscopic techniques. From these techniques, we had rationalized the interaction of 9-AA with various dyes via ground state complex formation. The binding constant (K) and number of binding sites (n) were calculated from the relevant fluorescence quenching data. The fluorescence emission of 9-Aminoacridine was quenched by the dyes through static quenching followed by the mechanism of energy transfer. The static quenching mechanism was confirmed by lifetime measurements. The thermodynamic parameters such as free energy change (ΔG), enthalpy change (ΔH) and entropy change (ΔS), were calculated by thermodynamic equations. The results showed that Van der Waals interaction or hydrogen bond formation played a major role in the binding of 9-AA with these dyes.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
9-Aminoacridine, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC)