Merck
  • Cu(OAc)2/TFA-promoted formal [3 + 3] cycloaddition/oxidation of enamines and enones for synthesis of multisubstituted aromatic amines.
Cu(OAc)2/TFA-promoted formal [3 + 3] cycloaddition/oxidation of enamines and enones for synthesis of multisubstituted aromatic amines.
Organic letters (2012-06-26)
Liang Li, Mi-Na Zhao, Zhi-Hui Ren, Jian-Li Li, Zheng-Hui Guan
RESUMEN
New strategies for the oxidative cycloaddition of enones with enamines are developed. These cycloaddition reactions directly afford substituted aromatic amines, which are important in organic chemistry, in moderate to good yield. Cu(OAc)(2)/TFA is shown to be essential to achieve high reaction efficiency.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Copper(II) acetate, 98%
Sigma-Aldrich
Copper(II) acetate, powder, 99.99% trace metals basis