Merck
  • Design, synthesis, biological evaluation and molecular modeling of novel 1,3,4-oxadiazole derivatives based on Vanillic acid as potential immunosuppressive agents.
Design, synthesis, biological evaluation and molecular modeling of novel 1,3,4-oxadiazole derivatives based on Vanillic acid as potential immunosuppressive agents.
Bioorganic & medicinal chemistry (2012-06-26)
Jian-Feng Tang, Xian-Hai Lv, Xiao-Liang Wang, Jian Sun, Yan-Bin Zhang, Yu-Shun Yang, Hai-Bin Gong, Hai-Liang Zhu
RESUMEN
In present study, a series of novel 1,3,4-oxadiazole derivatives have been designed, synthesized and purified. All of these compounds are reported for the first time, the chemical structures of these compounds were confirmed by means of (1)H NMR, ESI-MS and elemental analyses. Besides, we evaluated their immunosuppressive activity. Most of these synthesized compounds were proved to have potent immunosuppressive activity and low toxicity. Among them, the bioassay results demonstrated that compounds 5c, 5n, 5p, 5o, 6f and 6g exhibited immunosuppressive activities with IC(50) concentration range from 1.25μM to 7.60 μM against the T cells, and the IC(50) of positive control (csa) is 2.12 μM. Moreover, all the title compounds were assayed for PI3K/AKT signaling pathway inhibition using the ELISA assay. We examined the compounds with potent inhibitory activities against IL-1, IL-6 and IL-10 released in ConA-simulated mouse lymph node cells. The results showed compounds 5o and 6f displayed the most potential biological activity against T cells (IC(50)=1.25 μM and 4.75 μM for T cells). The preliminary mechanism of compound 5o inhibition effects was also detected by flow cytometry (FCM). The results of apoptosis and ELISA assay demonstrated that the immunosuppressive activity of compounds 5o and 6f against T cells may be mediated by the inhibition of PI3Kγ/AKT signaling pathway. Molecular docking was performed to position compounds 5o and 6f into PI3Kγ binding site in order to indicate the potential target.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Supelco
Vanillic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland