Merck
  • Development and characterization of an implantable biosensor for telemetric monitoring of ethanol in the brain of freely moving rats.
Development and characterization of an implantable biosensor for telemetric monitoring of ethanol in the brain of freely moving rats.
Analytical chemistry (2012-07-25)
Gaia Rocchitta, Ottavio Secchi, Maria Domenica Alvau, Rossana Migheli, Giammario Calia, Gianfranco Bazzu, Donatella Farina, Maria Speranza Desole, Robert D O'Neill, Pier Andrea Serra
RESUMEN
Ethanol is one of the most widespread psychotropic agents in western society. While its psychoactive effects are mainly associated with GABAergic and glutamatergic systems, the positive reinforcing properties of ethanol are related to activation of mesolimbic dopaminergic pathways resulting in a release of dopamine in the nucleus accumbens. Given these neurobiological implications, the detection of ethanol in brain extracellular fluid (ECF) is of great importance. In this study, we describe the development and characterization of an implantable biosensor for the amperometric detection of brain ethanol in real time. Ten different designs were characterized in vitro in terms of Michaelis-Menten kinetics (V(MAX) and K(M)), sensitivity (linear region slope, limit of detection (LOD), and limit of quantification (LOQ)), and electroactive interference blocking. The same parameters were monitored in selected designs up to 28 days after fabrication in order to quantify their stability. Finally, the best performing biosensor design was selected for implantation in the nucleus accumbens and coupled with a previously developed telemetric device for the real-time monitoring of ethanol in freely moving, untethered rats. Ethanol was then administered systemically to animals, either alone or in combination with ranitidine (an alcohol dehydrogenase inhibitor) while the biosensor signal was continuously recorded. The implanted biosensor, integrated in the low-cost telemetry system, was demonstrated to be a reliable device for the short-time monitoring of exogenous ethanol in brain ECF and represents a new generation of analytical tools for studying ethanol toxicokinetics and the effect of drugs on brain ethanol levels.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ranitidine hydrochloride, solid
Supelco
Ranitidine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material