Merck
Is there a nitrergic modulation of the rat external anal sphincter?
Experimental physiology (2012-08-09)
J Evers, M Buffini, S Craven, P R O'Connell, J F X Jones
RESUMEN
Nitric oxide is known to relax the internal anal sphincter, but its effect on the external anal sphincter (EAS) is unknown. The aim of this study was to investigate whether there is a nitrergic nerve plexus that modulates the EAS, similar to that found in oesophageal striated muscle. An in vitro ring preparation of rat anal canal was used to evaluate the effects of the nitric oxide synthase inhibitor N(ω)-nitro-L-arginine (L-NNA) and the NO donor sodium nitroprusside (SNP) on the EAS in conditions of neuromuscular blockade and the effect of SNP on nerve-evoked contractions. Immunohistological experiments were conducted to determine whether the neuronal isoform of nitric oxide synthase (nNOS) is present in the EAS. During direct muscle stimulation neither L-NNA (P = 0.32) nor SNP (P = 0.19) significantly changed the EF(50), which is the frequency at which 50% of maximal contraction is reached, compared with a time-dependent control. Nerve-evoked contractions were also not altered by addition of SNP to the tissue bath. Immunohistohistological experiments clearly showed co-localization of nNOS-positive nerve fibres at motor endplates of the oesophagus but not in the EAS. The internal anal sphincter was richly innervated by nitrergic fibres, but these did not extend into the EAS. In conclusion, there are no nitrergic motor fibres innervating the EAS, neurotransmission at the motor endplates is not affected by NO, and NO does not affect muscle force directly in conditions of neuromuscular blockade. There is, therefore, no evidence that EAS contraction is directly modulated by NO or by pudendal nitrergic fibres or diffusion from neighbouring nitrergic plexuses of the anal canal.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Seroalbúmina bovina, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Suero de cabra
Sigma-Aldrich
Nω-Nitro-L-arginine, ≥98% (TLC)
Sigma-Aldrich
IgG anti-conejo (molécula completa)-FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution