Merck
  • Kinetic modeling and occupancy measures of the norepinephrine transporters in baboons using single photon emission computed tomography with (123)I-INER.
Kinetic modeling and occupancy measures of the norepinephrine transporters in baboons using single photon emission computed tomography with (123)I-INER.
Synapse (New York, N.Y.) (2012-10-09)
Adriana Alexandre dos Santos Tavares, Olivier Barret, Jeffery Batis, David Alagille, Andrei Koren, Caroline Papin, Greg Kudej, Katie Nice, Jeffrey H Kordower, Kelly P Cosgrove, Tracy Kloczynski, Eric Brenner, John Seibyl, Gilles Tamagnan
RESUMEN
This study aims to investigate the pharmacokinetics of a recently developed radiotracer for imaging of the norepinephrine transporter (NET) in baboon brain, (123)I-INER, using single photon emission computed tomography (SPECT). In addition, it also aims to determine NET occupancy by atomoxetine and reboxetine, two selective norepinephrine reuptake inhibitors, using (123)I-INER in baboons. Baseline and preblocking studies with a high dose of atomoxetine (0.85 mg/kg) were conducted in three baboons using SPECT with (123)I-INER administered as a bolus. Kinetic modeling analysis was investigated for different models, namely invasive and reference tissue models. Bolus plus constant infusion experiments with displacement at equilibrium using six different doses of atomoxetine (0.03-0.85 mg/kg) and four different doses of reboxetine (0.5-3.0 mg/kg) were carried out in several baboons to obtain occupancy measurements as a function of dose for the two NET selective drugs. Results showed that reference tissue models can be used to estimate binding potential values and occupancy measures of (123)I-INER in different brain regions. In addition, the apparent volume of distribution was estimated by dividing concentration in tissue by the concentration in blood at 3 hours postinjection. After administration of atomoxetine or reboxetine, a dose-dependent occupancy was observed in brain regions known to contain high densities of NET. In conclusion, pharmacokinetic properties of (123) I-INER were successfully described, and obtained results may be used to simplify future data acquisition and image processing. Dose-dependent NET occupancy for two selective norepinephrine reuptake inhibitors was successfully measured in vivo in baboon brain using SPECT and (123) I-INER.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Atomoxetine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(R)-Tomoxetine hydrochloride, solid