Merck
  • Synthesis of a cholesteryl-HEG phosphoramidite derivative and its application to lipid-conjugates of the anti-HIV 5'TGGGAG³' Hotoda's sequence.
Synthesis of a cholesteryl-HEG phosphoramidite derivative and its application to lipid-conjugates of the anti-HIV 5'TGGGAG³' Hotoda's sequence.
Molecules (Basel, Switzerland) (2012-10-24)
Domenica Musumeci, Daniela Montesarchio
RESUMEN
A novel phosphoramidite derivative of cholesterol, with an ether-linked hexaethylene glycol (HEG) spacer arm, has been obtained through simple and reproducible solid phase modified oligonucleotide synthesis manipulations. This building block and the known phosphoramidite derivative of 3b-(2-hydroxyethoxy)cholesterol have been exploited in standard oligonucleotide synthesis protocols for the preparation of 5'- conjugates of the G-quadruplex-forming ⁵'TGGGAG³' oligomer, known as the Hotoda's sequence, to produce new potential anti-HIV agents.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Hexaethylene glycol, 97%