Merck
A carpal ligament substitute part II: polyester suture for scapho-lunate and triqueto-lunate ligament reconstruction.
Hand clinics (2012-11-22)
Marwan A Wehbé, Mary Lee Whitaker
RESUMEN
Carpal ligaments are commonly injured and may lead to pain and disability. These ligaments are very difficult to repair, and the results are unpredictable; as a result, treatment options abound. A novel approach is presented here using a polyester suture, aiming to substitute these ligaments' function, rather than to repair them.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Poly(ethylene terephthalate), granular