Merck
Early childhood caries: risk-based disease prevention and management.
Dental clinics of North America (2012-11-24)
Man Wai Ng, Isabelle Chase
RESUMEN
Early childhood caries (ECC), common in preschoolers, can lead to pain and infection if left untreated. Yet, ECC is largely preventable, and if it is identified early and the responsible risk factors are addressed, its progression can be halted or slowed. This article reviews the rationale for a first dental visit by age 1 year, caries risk assessment, and risk-based prevention and management of ECC and discusses strategies for providers to implement these contemporary evidence-based concepts into clinical practice.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Xylitol, ≥99% (GC)
Sigma-Aldrich
Xylitol
Supelco
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Xylitol, analytical standard