Merck
Non-oxidative ethanol metabolites as a measure of alcohol intake.
Clinica chimica acta; international journal of clinical chemistry (2012-11-28)
Thomas M Maenhout, Marc L De Buyzere, Joris R Delanghe
RESUMEN
Recent alcohol intake can be monitored by the measurement of indirect biomarkers. Elevated levels of liver enzymes (i.e. gamma-glutamyl transferase (GGT), alanine amino transferase (ALT) and aspartate amino transferase (AST)) in blood are commonly used in clinical practice as an indicator of alcohol-induced liver damage. With the exception of carbohydrate-deficient transferrin (CDT), the specificity of indirect markers is only moderate because many cases of elevated levels are unrelated to alcohol consumption. Because of their intermediate half-life and tendency to accumulate in hair, non-oxidative ethanol metabolites can be used as markers with an intermediate timeframe between ethanol measurements and GGT and CDT with regard to recent alcohol consumption occurring between hours to 1 week. Additionally, these biomarkers offer a high ethanol-specificity in combination with approximately a two-fold higher sensitivity in comparison with indirect alcohol markers. In case of forensic use of direct ethanol metabolites, caution has to be taken in interpretation and pre-analytical pitfalls should be considered.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Etil-β-D-glucurónido, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Diethyl sulfate, 98%
Supelco
Etil-β-D-glucurónido, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®