Merck
[Chemical constituents from Imperata cylindrica].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-11-30)
Xuan Liu, Binfeng Zhang, Guixin Chou, Li Yang, Zhengtao Wang
RESUMEN
Chemical investigation of Imperata cylindrica led to the isolation of thirteen compounds using various chromatographic techniques. The structure of these compounds were identified as: three phenylpropanoids, 1-(3,4,5-trimethoxyphenyl)-1,2,3-propanetriol ( 1 ), 1-O-p-coumaroylglycerol (2), 4-methoxy-5-methyl coumarin-7-O-beta-D-glucopyranoside (3); four organic acids, 4-hydroxybenzene carboxylic acid(4), 3,4-dihydroxybenzoic acid (5), vanillic acid (6), 3, 4-dihydroxybutyric acid (7); one phenolic compound, salicin (8); and five triterpenes, namely, arundoin (9), cylindrin (10), fernenol (11), simiarenol (12), glutinone (13) by their physicochemical properties and spectral data analysis. Among them, compounds 1-8 were isolated from the genus Imperata for the first time.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Protocatechuic acid, primary reference standard
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Supelco
3,4-Dihydroxybenzoic acid, analytical standard
Supelco
Vanillic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland