Merck
Peptide/protein separation with cationic polymer brush nanosponges for MALDI-MS analysis.
Langmuir : the ACS journal of surfaces and colloids (2012-12-19)
Bojan Mitrovic, Stephanie Eastwood, VenNey Wong, Daniel Dyer, Gary Kinsel, Colleen Scott
RESUMEN
A cationic polymer nanobrush was synthesized, attached to a MALDI target, and used for the fractionation of peptides and proteins based on their pI, prior to analysis by MALDI-MS. The cationic polymer nanobrush was synthesized on a gold substrate by AIBN photoinitiated polymerization, using a 70:30 ratio of 2-aminoethyl methacrylate hydrochloride (AEMA):N-isopropylacrylamide (NIPAAM). This brush showed selectivity for adsorption of acidic peptides and proteins and allowed fractionation of simple two-component mixtures to be completed in less than 10 min. The brush-adsorbed biomolecules were recovered by treating the nanobrush with ammonium hydroxide, which effectively collapsed the brush, thereby releasing the trapped compounds for MALDI MS analysis. These results demonstrate that nanobrush can serve as a convenient platform for rapid fractionation of biomolecules prior to analysis by MALDI-MS.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Hidróxido de amonio solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Hidróxido de amonio solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
N-Isopropylacrylamide, 97%
Sigma-Aldrich
N-Isopropylacrylamide, ≥99%
Sigma-Aldrich
Hidróxido de amonio solution, BioUltra, ~1 M NH3 in H2O (T)