Merck
Rheological behavior of xanthan gum solution related to shear thinning fluid delivery for subsurface remediation.
Journal of hazardous materials (2012-12-19)
L Zhong, M Oostrom, M J Truex, V R Vermeul, J E Szecsody
RESUMEN
Xanthan gum solutions are shear thinning fluids which can be used as delivery media to improve the distribution of remedial amendments injected into heterogeneous subsurface environments. The rheological behavior of the shear thinning solution needs to be known to develop an appropriate design for field injection. In this study, the rheological properties of xanthan gum solutions were obtained under various chemical and environmental conditions relevant to delivery of remedial amendments to groundwater. Higher xanthan concentration raised the absolute solution viscosity and increased the degree of shear thinning. Addition of remedial amendments (e.g., phosphate, sodium lactate, ethyl lactate) caused the dynamic viscosity of xanthan solutions to decrease, but they maintained shear-thinning properties. Use of mono- and divalent salts (e.g., Na(+), Ca(2+)) to increase the solution ionic strength also decreased the dynamic viscosity of xanthan and the degree of shear thinning, although the effect reversed at high xanthan concentrations. A power law analysis showed that the consistency index is a linear function of the xanthan concentration. The degree of shear thinning, however, is best described using a logarithmic function. Mechanisms to describe the observed empiricism have been discussed. In the absence of sediments, xanthan solutions maintained their viscosity for months. However, the solutions lost their viscosity over a period of days to weeks when in contact with site sediment. Loss of viscosity is attributed to physical and biodegradation processes.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Fosfato de sodio dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Fosfato de sodio dibasic heptahidratado, ACS reagent, 98.0-102.0%
Sigma-Aldrich
Cloruro de calcio dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Fosfato de sodio dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Cloruro de calcio dihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Cloruro de calcio solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Fosfato de sodio dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Fosfato de sodio dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Cloruro de calcio dihydrate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Cloruro de calcio dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Fosfato de sodio dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Fosfato de sodio dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Fosfato de sodio dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Fosfato de sodio dibasic, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
Fosfato de sodio dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium DL-lactate solution, syrup, BioReagent, 60 % (w/w), synthetic, suitable for cell culture
Sigma-Aldrich
Ethyl lactate, ≥98%, FCC, FG
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
(−)-Ethyl L-lactate, 98%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)