Merck
Synthesis, characterization and preliminary anticonvulsant evaluation of some 4-alkyl-1,2,4-triazoles.
European journal of medicinal chemistry (2013-01-08)
Tomasz Plech, Jarogniew J Luszczki, Monika Wujec, Jolanta Flieger, Magdalena Pizoń
RESUMEN
Designed and synthesized 4-alkyl-1,2,4-triazole-3-thione derivatives showed significant anticonvulsant activity, determined in the maximal electroshock-induced seizure (MES) test. The chemical structure of all new compounds was confirmed by spectral methods ((1)H NMR, (13)C NMR, IR, MS). A sensitive and selective method was elaborated for the determination of the anticonvulsant compounds levels in mice brain tissue, based on HPLC with diode array detector (DAD). Chromatographic tests showed that lack of anticonvulsant effect of two derivatives (15, 16) with long alkyl chains at N-4 position of the 1,2,4-triazole ring was due to the inability to cross the blood-brain barrier (BBB).
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
1,2,4-Triazole, 98%