Merck
Total synthesis of (±)-cavicularin: control of pyrone Diels-Alder regiochemistry using isomeric vinyl sulfones.
Organic letters (2013-01-11)
Peng Zhao, Christopher M Beaudry
RESUMEN
An intramolecular pyrone Diels-Alder reaction-elimination retro-Diels-Alder cascade of a vinyl sulfone was used in the synthesis of cavicularin, a molecule possessing conformational chirality. The vinyl sulfone substitution pattern allowed for regiocontrol in the Diels-Alder cascade event.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Divinyl sulfone, contains hydroquinone as inhibitor, ≥96%