Merck
Detection of some volatile degradation products released during photoexposition of ranitidine in a solid state.
Journal of pharmaceutical and biomedical analysis (2013-01-22)
Marzena Jamrógiewicz, Bartosz Wielgomas
RESUMEN
Ranitidine (RAN) is on top of the list of prescribed drugs, due to its popularity as a selective H2-receptor antagonist, which efficiently decreases the amount of acid produced in the stomach. RAN is not stable both in a solid state and in a solution, which creates manufacturing problems, requires appropriate storage conditions, and results in a short drug shelf-life. The aim of this work was to study the emission of volatile degradation products generated during photoexposition of ranitidine hydrochloride in a solid state. Significant changes in volatile profile of irradiated RAN were detected using HS-SPME-GC-MS. Sixteen major peaks were noticed on the chromatograms of irradiated ranitidine and the structures of some compounds were elucidated, while the presence of acetaldoxime, thiazole, dimethylformamide, dimethylacetamide and 5-methylfurfural was confirmed by means of the analysis of the authentic standards.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
5-Methylfurfural, ReagentPlus®, 99%
Sigma-Aldrich
Ranitidine hydrochloride, solid
Supelco
Ranitidine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material