Merck
[Effects of plant growth regulator mepiquat chloride on content of active compounds in Scutellaria baicalensis].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-02-13)
Guo-Qiang Hu, Xue-Wen Zhang, Min-Hui Li, Guo-Hu Song, Yuan Yuan, Shu-Fang Lin, Zhi-Gang Wu
RESUMEN
To study the effect of plant growth regulator mepiquat chloride (DPC) on the growing development of plant and the content of active compounds in Scutellaria baicalensis. After spraying DPC during the seedling period, the length of taproot, fresh weight of root, diameter of taproot and the length of stem were measured. The contents of baicalin, baicalein and wogonin were determined by HPLC, respectively. Total flavonoids and scavenging DPPH were determined with ultraviolet spectrophotometry. After spraying DPC, fresh weight of S. baicalensis root was significant increased, and root diameter was also increased. The contents of baicalin and total flavonoids were significantly increased and baicalein and wogonin were decreased remarkably. Compared with controls, scavenging activity of ethanol extracts on DPPH free radical was no significant changed after spraying DPC. Plant growth regulator DPC could regulate the growth on the ground and underground effectively, and could enhance the content of flavonoids compounds of S. baicalensis.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Mepiquat chloride, PESTANAL®, analytical standard