Merck
Droplet-based microfluidic system to form and separate multicellular spheroids using magnetic nanoparticles.
Lab on a chip (2013-02-22)
Sungjun Yoon, Jeong Ah Kim, Seung Hwan Lee, Minsoo Kim, Tai Hyun Park
RESUMEN
The importance of creating a three-dimensional (3-D) multicellular spheroid has recently been gaining attention due to the limitations of monolayer cell culture to precisely mimic in vivo structure and cellular interactions. Due to this emerging interest, researchers have utilized new tools, such as microfluidic devices, that allow high-throughput and precise size control to produce multicellular spheroids. We have developed a droplet-based microfluidic system that can encapsulate both cells and magnetic nanoparticles within alginate beads to mimic the function of a multicellular tumor spheroid. Cells were entrapped within the alginate beads along with magnetic nanoparticles, and the beads of a relatively uniform size (diameters of 85% of the beads were 170-190 μm) were formed in the oil phase. These beads were passed through parallel streamlines of oil and culture medium, where the beads were magnetically transferred into the medium phase from the oil phase using an external magnetic force. This microfluidic chip eliminates additional steps for collecting the spheroids from the oil phase and transferring them to culture medium. Ultimately, the overall spheroid formation process can be achieved on a single microchip.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Alginic acid sodium salt, powder
Sigma-Aldrich
Alginato de sodio
Sigma-Aldrich
Ácido algínico sodium salt from brown algae, Medium viscosity
Sigma-Aldrich
Ácido algínico sodium salt from brown algae, low viscosity
Sigma-Aldrich
Ácido algínico sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Alginic acid from brown algae, powder
Sigma-Aldrich
Alginic acid calcium salt from brown algae