Merck
  • A novel melittin-MhIL-2 fusion protein inhibits the growth of human ovarian cancer SKOV3 cells in vitro and in vivo tumor growth.
A novel melittin-MhIL-2 fusion protein inhibits the growth of human ovarian cancer SKOV3 cells in vitro and in vivo tumor growth.
Cancer immunology, immunotherapy : CII (2013-02-28)
Mingjun Liu, Jinbao Zong, Zimin Liu, Ling Li, Xu Zheng, Bin Wang, Guirong Sun
RESUMEN
In the current study, we produced a novel fusion protein (melittin-mutant human interleukin 2, melittin-MhIL-2) comprising a mutant human interleukin 2 (Arg88/Ala125) genetically linked to melittin. The plasmid pET15b-melittin-MhIL-2 (Arg88/Ala125) was transformed into E. coli for protein expression. The expressed melittin-MhIL-2 protein was purified using a series of purification steps. The interleukin 2 (IL-2) activity of melittin-MhIL-2 fusion protein was compared with recombinant human interleukin 2 (rhIL-2) for its ability to induce CTLL-2 proliferation. Moreover, the fusion protein directly inhibits the growth of human ovarian cancer SKOV3 cells in vitro. In an in vivo initial experiment, the fusion protein inhibited tumor growth in ovarian cancer mice. In conclusion, we generated a novel melittin-MhIL-2 fusion protein that retained functional activity of IL-2 and melittin and inhibited tumor growth in vivo.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Melittin from honey bee venom, ≥85% (HPLC)
Sigma-Aldrich
Melittin, ≥97% (HPLC), synthetic
Sigma-Aldrich
Melittin from honey bee venom, ≥65% (HPLC)