Merck
Synergistic insertion of antimicrobial magainin-family peptides in membranes depends on the lipid spontaneous curvature.
Biophysical journal (2013-03-27)
Erik Strandberg, Jonathan Zerweck, Parvesh Wadhwani, Anne S Ulrich
RESUMEN
PGLa and magainin 2 (MAG2) are amphiphilic antimicrobial peptides from frog skin with known synergistic activity. The orientation of the two helices in membranes was studied using solid-state (15)N-NMR, for each peptide alone and for a 1:1 mixture of the peptides, in a range of different lipid systems. Two types of orientational behavior emerged. 1), In lipids with negative spontaneous curvature, both peptides remain flat on the membrane surface, when assessed both alone and in a 1:1 mixture. 2), In lipids with positive spontaneous curvature, PGLa alone assumes a tilted orientation but inserts into the bilayer in a transmembrane alignment in the presence of MAG2, whereas MAG2 stays on the surface or gets only slightly tilted, when observed both alone and in the presence of PGLa. The behavior of PGLa alone is identical to that of another antimicrobial peptide, MSI-103, in the same lipid systems, indicating that the curvature-dependent helix orientation is a general feature of membrane-bound peptides and also influences their synergistic intermolecular interactions.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Magainin II, ≥97% (HPLC)