Merck
Identification and treatment of amblyopia.
American family physician (2013-04-04)
Yasmin S Bradfield
RESUMEN
Amblyopia is the leading cause of vision loss in children. It is treatable if diagnosed early, making identification of affected children critical. The American Association for Pediatric Ophthalmology and Strabismus and the American Academy of Pediatrics recommend that clinicians routinely perform age-appropriate vision chart testing, red reflex testing, and examination for signs of strabismus. The U.S. Preventive Services Task Force recommends vision screening for all children at least once between three and five years of age to detect the presence of amblyopia or its risk factors. Photoscreening may be a useful adjunct to traditional vision screening, but there is limited evidence that it improves visual outcomes. Treatments for amblyopia include patching, atropine eye drops, and optical penalization of the nonamblyopic eye. In children with moderate amblyopia, patching for two hours daily is as effective as patching for six hours daily, and daily atropine is as effective as daily patching. Children older than seven years may still benefit from patching or atropine, particularly if they have not previously received amblyopia treatment. Amblyopia recurs in 25 percent of children after patching is discontinued. Tapering the amount of time a patch is worn each day at the end of treatment reduces the risk of recurrence.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Atropine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Atropine for peak identification, European Pharmacopoeia (EP) Reference Standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad