Merck
Sequestration of a highly reactive intermediate in an evolving pathway for degradation of pentachlorophenol.
Proceedings of the National Academy of Sciences of the United States of America (2013-05-17)
Itamar Yadid, Johannes Rudolph, Klara Hlouchova, Shelley D Copley
RESUMEN
Microbes in contaminated environments often evolve new metabolic pathways for detoxification or degradation of pollutants. In some cases, intermediates in newly evolved pathways are more toxic than the initial compound. The initial step in the degradation of pentachlorophenol by Sphingobium chlorophenolicum generates a particularly reactive intermediate; tetrachlorobenzoquinone (TCBQ) is a potent alkylating agent that reacts with cellular thiols at a diffusion-controlled rate. TCBQ reductase (PcpD), an FMN- and NADH-dependent reductase, catalyzes the reduction of TCBQ to tetrachlorohydroquinone. In the presence of PcpD, TCBQ formed by pentachlorophenol hydroxylase (PcpB) is sequestered until it is reduced to the less toxic tetrachlorohydroquinone, protecting the bacterium from the toxic effects of TCBQ and maintaining flux through the pathway. The toxicity of TCBQ may have exerted selective pressure to maintain slow turnover of PcpB (0.02 s(-1)) so that a transient interaction between PcpB and PcpD can occur before TCBQ is released from the active site of PcpB.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cóctel de inhibidores de proteasas, for use in purification of Histidine-tagged proteins, DMSO solution
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina hydrate, ≥95% (HPLC)
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina hydrate, ≥99%
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Tetrachloro-1,4-benzoquinone, 99%
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina hydrate, Grade AA-1
Sigma-Aldrich
5′-Monofosfato de riboflavina sodium salt hydrate, 70-79% (fluorimetric)
Sigma-Aldrich
5′-Monofosfato de riboflavina sodium salt hydrate, synthetic, ≥70% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
Pentachlorophenol, 97%
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Riboflavin sodium phosphate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina hydrate, purified by column chromatography, ≥99%
Supelco
Pentachlorophenol solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
5′-Monofosfato de riboflavina sodium salt hydrate, suitable for electrophoresis, suitable for acrylamide photopolymerization, ≥70%, powder
Sigma-Aldrich
3,4,5,6-Tetrachloro-1,2-benzoquinone, 97%
Sigma-Aldrich
Riboflavin Sodium Phosphate Hydrate, tested according to Ph. Eur.
Supelco
Pentachlorophenol, analytical standard