Merck
  • Tryptic digestion of bovine secretory IgA at elevated temperature and in urea. Isolation of SC domain 1 which is covalently bound to IgA dimer and binds non-covalently to IgM.
Tryptic digestion of bovine secretory IgA at elevated temperature and in urea. Isolation of SC domain 1 which is covalently bound to IgA dimer and binds non-covalently to IgM.
The International journal of biochemistry (1989-01-01)
D Beale
RESUMEN
1. Tryptic cleavage sites in bovine secretory component (SC) which become inaccessible when SC is bound to IgA dimer remained inaccessible at 60 degrees C and in 4 M urea at 37 degrees C. 2. This suggests the presence of strong interactions compatible with published affinity constants of ca 10(8) M-1. 3. In 5 M urea at 37 degrees C further cleavage of bound SC did occur to produce a fragment consisting of domain 1 which was disulphide bridged to the IgA dimer. 4. Binding studies on the isolated fragment showed that domain 1 did not account for all the binding by SC. 5. Cleavage of the isolated fragment with iodosobenzoic produced a smaller fragment consisting of the n-terminal third of domain 1 (residues 1-35). This N-terminal fragment showed significant binding.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
2-Iodosobenzoic acid