Merck
Anthraflavic acid inhibits the mutagenicity of the food mutagen IQ: mechanism of action.
Mutation research (1988-03-01)
A D Ayrton, C Ioannides, R Walker
RESUMEN
The ability of anthraflavic acid to inhibit the mutagenicity of IQ was investigated using the Ames test and employing hepatic activation systems from Aroclor 1254-pretreated rats. Incorporation of anthraflavic acid into the S9 mix caused a concentration-dependent decrease in the mutagenicity of IQ. A similar effect was seen when microsomes only were employed as activation systems. Cytosol, as we have previously demonstrated, potentiated the microsome-mediated mutagenicity of IQ and this potentiation was also inhibited by anthraflavic acid. In contrast, anthraflavic acid had no effect on the mutagenicity of the direct-acting microsome-generated metabolites of IQ. It is concluded that anthraflavic acid is a potent inhibitor of IQ mutagenicity by virtue of its ability to inhibit both its microsomal and cytosolic activation pathways.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anthraflavic acid, technical grade, 90%