Merck
Genotoxicity of carcinogenic N-nitrosopropylamine derivatives in the hepatocyte primary culture/DNA-repair test.
Mutation research (1985-11-01)
H Yamazaki, Y Mori, K Toyoshi, H Mori, S Sugie, N Yoshimi, Y Konishi
RESUMEN
The genotoxicity of N-nitrosodipropylamine, 8 of its oxidized derivatives and N-nitroso-2,6-dimethylmorpholine was examined in the hepatocyte primary culture (HPC)/DNA repair test. Nine N-nitrosamines which are known to be carcinogenic and mutagenic were clearly positive in the HPC/DNA-repair test. N-Nitroso(2,3-dihydroxypropyl) (2-hydroxypropyl)amine did not elicit DNA repair, but showed a borderline mutagenic response in the Salmonella/microsome test. Thus, the HPC/DNA-repair test displays a comparable capacity to the bacterial mutagenesis test for detecting the genotoxic effects of this class of carcinogens.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
N-Nitrosodi-n-propylamine, analytical standard
Supelco
N-Nitrosodi-n-propylamine solution, certified reference material, 5000 μg/mL in methanol