Merck
  • Isopycnic-centrifugation studies in caesium chloride and in caesium sulphate on dermatan sulphate proteoglycans from bovine sclera.
Isopycnic-centrifugation studies in caesium chloride and in caesium sulphate on dermatan sulphate proteoglycans from bovine sclera.
The Biochemical journal (1981-12-01)
J K Sheehan, I Carlstedt, L Cöster, A Malmström, L A Fransson
RESUMEN
1. Two proteodermatan sulphate species from bovine sclera (fractions PG-I and PG-II) separable by gel chromatography were studied by isopycnic centrifugations in CsCl and Cs(2)SO(4) both in an analytical and a preparative mode. 2. In CsCl, fraction PG-I formed a broad band at a density rho=1.75g/ml whereas fraction PG-II banded sharply at rho=1.64g/ml. However, in Cs(2)SO(4), fraction PG-II banded at rho=1.51g/ml and fraction PG-I at rho=1.40g/ml, a reversal of the relative banding positions of the two species in CsCl. 3. Preparative isopycnic centrifugations in the two caesium salts permitted further subfractionation of fractions PG-I and PG-II. In both CsCl and Cs(2)SO(4) fraction PG-I was split into subfractions that varied greatly in uronate/protein ratios but had very similar uronate composition. In contrast, isopycnic centrifugation of fraction PG-II in Cs(2)SO(4) gave rise to subfractions with similar uronate/protein ratios but markedly different uronate composition (iduronate content, 88-42%). 4. Subfractions of fractions PG-I and PG-II obtained in preparative centrifugations in CsCl or Cs(2)SO(4) were examined in the analytical ultracentrifuge. These subfractions banded at discrete positions in the gradient. Estimations of apparent molecular weight for these subfractions from data from the analytical isopycnic centrifugations gave values that were much higher (around 1x10(6)) than were those obtained previously for their ;monomeric' states (fraction PG-I 160000-410000, and fraction PG-II 70000-130000). 5. In CsCl, fraction PG-I may be subfractionated according to the number of side chains in the molecule. Fraction PG-I increased its net solvation (i.e. lowered its buoyant density) to a larger extent than did fraction PG-II in going from CsCl to Cs(2)SO(4) (i.e. from lower to higher water activity). It is proposed that the presence of large amounts of iduronate in fraction PG-II makes these molecules relatively less solvated in Cs(2)SO(4). Thus the uronate composition may be an important factor in determining the banding position of proteodermatan sulphates in density-gradient centrifugations.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cesium sulfate, Grade I, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Cesium sulfate, 99.99% trace metals basis
En este momento no podemos mostrarle ni los precios ni la disponibilidad