Merck
  • The determination of cyclophosphamide and its metabolites in blood plasma as stable trifluoroacetyl derivatives by electron capture chemical ionization gas chromatography/mass spectrometry.
The determination of cyclophosphamide and its metabolites in blood plasma as stable trifluoroacetyl derivatives by electron capture chemical ionization gas chromatography/mass spectrometry.
Biological mass spectrometry (1994-03-01)
G Momerency, K Van Cauwenberghe, P H Slee, A T Van Oosterom, E A De Bruijn
RESUMEN
A method is described for the determination of the antitumour drug cyclophosphamide and six stable metabolites in plasma of cancer patients, namely dechloroethyl-cyclophosphamide, 4-keto-cyclophosphamide, carboxy-phosphamide, alcophosphamide, nor-nitrogen mustard and the N-chloroethyl-1,3-oxazolidine-2-one, as methyl and/or trifluoroacetyl derivatives by single ion monitoring gas chromatography/mass spectrometry, mostly in the electron capture chemical ionization mode. The isolation of most metabolites was performed by solid-phase C-18 extraction in weakly acidic medium. The phosphoramide mustard isolated under these conditions decomposes readily to the nor-nitrogen mustard during derivatization. The original nor-nitrogen mustard and the chloroethyl-1,3-oxazolidine-2-one were isolated by liquid extraction with ethyl acetate in alkaline medium. Recoveries of 75-99% were measured using spiked blank plasma samples. Quantitation of metabolites in patient plasma samples was performed using two sets of calibration curves for the concentration ranges of 1-100 ng and 0.1-10 micrograms of metabolite per millilitre of original plasma.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Bis(2-chloroethyl)amine hydrochloride, 98%