Merck
  • Thiazolidine diones, specific ligands of the nuclear receptor retinoid Z receptor/retinoid acid receptor-related orphan receptor alpha with potent antiarthritic activity.
Thiazolidine diones, specific ligands of the nuclear receptor retinoid Z receptor/retinoid acid receptor-related orphan receptor alpha with potent antiarthritic activity.
The Journal of biological chemistry (1996-06-07)
M Missbach, B Jagher, I Sigg, S Nayeri, C Carlberg, I Wiesenberg
RESUMEN
Rat adjuvant arthritis is a chronic T cell-dependent autoimmune disease with many similarities to rheumatoid arthritis. We have identified a class of thiazolidine diones with high potency in suppressing chronic inflammation and joint destruction in this experimental model. The lead compound CGP 52608 (1-(3-allyl-4-oxothiazolidine-2-ylidene)-4-methylthiosemicarbazone) exhibits antiarthritic activity at daily oral doses between 0.01 and 1 mg/kg and was shown to specifically activate the retinoid Z receptor/retinoid acid receptor-related orphan receptor alpha (RZR/RORalpha) in low nanomolar concentrations. This receptor is a novel member of the superfamily of ligand-inducible transcription factors, and we have recently identified the pineal gland hormone melatonin as a natural ligand. Structure-activity relationship studies with 13 closely related analogues of CGP 52608 revealed a striking correlation between RZR/RORalpha activation and antiarthritic activity. We therefore suggest that nuclear signaling via RZR/RORalpha is a key mechanism in mediating the antiarthritic effects of these thiazolidine diones and may open a novel therapeutic approach for the treatment of rheumatoid arthritis and other autoimmune diseases. The existence of a nuclear melatonin receptor may lead to a better understanding of the immunomodulatory actions of melatonin.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
CGP 52608, ≥98%, solid