Merck
Functional expression of the human MDR1 gene in Escherichia coli.
Archives of biochemistry and biophysics (1996-09-01)
A M George, M W Davey, A A Mir
RESUMEN
In this preliminary study, we report the cloning of the human MDR1 cDNA into a prokaryotic expression vector and the consequent functional expression of heterologous P-glycoprotein in Escherichia coli. We demonstrate increased resistance to the P-glycoprotein substrates TPA+, TPP+, and puromycin; reduced accumulation of TPP+ and tetracycline by resistant cells; and the expression of a full-length immunoreactive P-glycoprotein molecule in the membrane fraction of resistant cells. The obvious structural and functional similarities of P-gp to prokaryotic ABC transporters and other efflux transporters argues for a more complete study of the consequences pertaining to the expression of human P-glycoprotein in E. coli.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Tetraphenylarsonium(V) chloride hydrate, 97%