Merck
  • Crystal structure and solid-state 13C NMR analysis of N-o-, N-m- and N-p-nitrophenyl-2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosylamines, and their N-acetyl derivatives.
Crystal structure and solid-state 13C NMR analysis of N-o-, N-m- and N-p-nitrophenyl-2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosylamines, and their N-acetyl derivatives.
Carbohydrate research (2006-09-05)
Andrzej Temeriusz, Tomasz Gubica, Paulina Rogowska, Katarzyna Paradowska, Michał K Cyrański
RESUMEN
The X-ray diffraction analysis of N-o-nitrophenyl-2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosylamine (1), N-m-nitrophenyl-2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosylamines, N-p-nitrophenyl-2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosylamines, and their N-acetyl derivatives was performed. The sugar moieties always adopt (4)C1 conformations, however, due to crystal packing forces they are always slightly distorted. It was found that except N-acetyl, N-m-nitrophenyl-2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosylamine (5), none of the glucopyranosylamines studied in this paper form strong hydrogen bonds in the crystal lattice. Additionally, (5) crystallizes with a molecule of water, which occupies a special crystallographic position (on the twofold axis) and links two sugar molecules by hydrogen bonds. The CP MAS NMR spectra confirmed the presence of the intermolecular hydrogen bond involving the molecule of water in (5). Moreover, it was proved that in (1) an intramolecular hydrogen bond is formed between the glycosidic linkage and the nitro group.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
4-Nitrophenyl N-acetyl-β-D-glucosaminide, ≥99% (TLC)
Sigma-Aldrich
4-Nitrophenyl N-acetyl-α-D-glucosaminide, ≥98%