Merck
Kinetics studies on triacontanyl palmitate: a urease inhibitor.
Natural product research (2007-07-10)
Muhammad Arif Lodhi, Muhammad Athar Abbasi, Muhammad Iqbal Choudhary, Viqar Uddin Ahmad
RESUMEN
The mechanism of inhibition of jack bean and Bacillus pasteurii ureases was investigated by triacontanyl palmitate (1) which is a long-chain fatty ester and has been isolated from Symplocos racemosa Roxb. Lineweaver-Burk, Dixon plots, and their secondary replots showed that 1 is a non-competitive inhibitor of these enzymes. K(i) values were found to be 60.03 +/- 1.72 and 88.23 +/- 0.31 microM against jack bean and B. pasteurii ureases, respectively.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
1-Triacontanol, ≥98% (capillary GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad