Merck
Basic fibroblast growth factor increases intracellular magnesium concentration through the specific signaling pathways.
Molecules and cells (2009-08-28)
Bing-Zhe Hong, Sun-Ah Park, Han-Na Kim, Tian-Ze Ma, Han-Gyu Kim, Hyung-Sub Kang, Hwan-Gyu Kim, Yong-Geun Kwak
RESUMEN
Basic fibroblast growth factor (bFGF) plays an important role in angiogenesis. However, the underlying mechanisms are not clear. Mg(2+) is the most abundant intracellular divalent cation in the body and plays critical roles in many cell functions. We investigated the effect of bFGF on the intracellular Mg(2+) concentration ([Mg(2+)](i)) in human umbilical vein endothelial cells (HUVECs). bFGF increased [Mg(2+)](i) in a dose-dependent manner, independent of extracellular Mg(2+). This bFGF-induced [Mg(2+)](i) increase was blocked by tyrosine kinase inhibitors (tyrphostin A-23 and genistein), phosphatidylinositol 3-kinase (PI3K) inhibitors (wortmannin and LY294002) and a phospholipase Cgamma (PLCgamma) inhibitor (U73122). In contrast, mitogen-activated protein kinase inhibitors (SB202190 and PD98059) did not affect the bFGF-induced [Mg(2+)](i) increase. These results suggest that bFGF increases the [Mg(2+)](i) from the intracellular Mg(2+) stores through the tyrosine kinase/PI3K/PLCgamma-dependent signaling pathways.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Tyrphostin 23, ≥98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad