Merck
  • Molecular structural studies of lichen substances with antimicrobial, antiproliferative, and cytotoxic effects from Parmelia subrudecta.
Molecular structural studies of lichen substances with antimicrobial, antiproliferative, and cytotoxic effects from Parmelia subrudecta.
Preparative biochemistry & biotechnology (2010-11-26)
Veneta Ivanova, Martin Backor, Hans-Martin Dahse, Udo Graefe
RESUMEN
Lecanoric acid (1), orsellinic acid methyl ester (2), orcinol (3), and usnic acid (4) were isolated from the lichen Parmelia subrudecta, collected on Palma of the Canary Islands, Spain. Compounds 1, 2, 3, and 4 were purified by solvent extraction, silica gel column chromatography, and preparative high-performance liquid chromatography (HPLC) consecutively. The structures of the four compounds were elucidated by one- and two-dimensional nuclear magnetic resonance (NMR) experiments and mass spectrometric investigations. These compounds showed activity against important gram-positive and gram-negative pathogens like mycobacteria and multiresistant staphylococci. This activity is combined with antiproliferative activity and cytotoxicity.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Orcinol, 97%