Merck
Fructose-1,6-bisphosphate protects against Zymosan-induced acute lung injury in mice.
Inflammation (2012-02-14)
Roberto Christ Vianna Santos, Rafael Noal Moresco, Miguel Angel Peña Rico, Antonio R García Susperregui, Jose Luis Rosa, Ramon Bartrons, Francesc Ventura, Débora Nunes Mário, Sydney Hartz Alves, Etiane Tatsch, Helena Kober, Ricardo Obalski de Mello, Patrícia Scherer, Jarbas Rodrigues de Oliveira
RESUMEN
It has been previously showed that fructose-1,6-bisphosphate (FBP) has anti-inflammatory and immunomodulatory effects on several experimental inflammation models. However, the effects and mechanism of FBP on Zymosan-induced acute lung injury (ALI) in mice had not been tested. In this study, our aim was to assess the anti-inflammatory activities of FBP on Zymosan-induced ALI. We found that in vivo treatment with FBP (500 mg/kg i.p.) markedly decreased the nitric oxide (NO) levels in the lungs and significantly reduced bronchoalveolar lavage fluid total cell and neutrophil counts and protein exudation after Zymosan challenge. Furthermore, FBP inhibited inducible nitric oxide synthase (iNOS) activities in RAW macrophages. Meanwhile, FBP did not inhibit the cyclooxigenase 2, interleukin-6, and nuclear factor kappa B transcription. Taken together, these results suggest that FBP shows anti-inflammatory effects through inhibiting lung edema, NO, and iNOS activities.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
D-Fructosa 1,6-bisfosfato trisodium salt hydrate, ≥98% (TLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad