Merck
Difluoro(trimethylsilyl)acetonitrile: synthesis and fluoroalkylation reactions.
The Journal of organic chemistry (2012-06-20)
Mikhail D Kosobokov, Alexander D Dilman, Vitalij V Levin, Marina I Struchkova
RESUMEN
A new silicon reagent, difluoro(trimethylsilyl)acetonitrile, was prepared by insertion of difluorocarbene into silyl cyanide. The obtained silane served as a good cyanodifluoromethylating reagent toward aldehydes, N-tosylimines, N-alkylimines, and enamines under basic or acidic conditions.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Trimethylsilyl cyanide, 98%
Sigma-Aldrich
Trimethylsilyl cyanide, technical, ≥95% (GC)