Merck
  • Scalable syntheses of both enantiomers of DNJNAc and DGJNAc from glucuronolactone: the effect of N-alkylation on hexosaminidase inhibition.
Scalable syntheses of both enantiomers of DNJNAc and DGJNAc from glucuronolactone: the effect of N-alkylation on hexosaminidase inhibition.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-06-28)
Andreas F G Glawar, Daniel Best, Benjamin J Ayers, Saori Miyauchi, Shinpei Nakagawa, Matilde Aguilar-Moncayo, José M García Fernández, Carmen Ortiz Mellet, Elizabeth V Crabtree, Terry D Butters, Francis X Wilson, Atsushi Kato, George W J Fleet
RESUMEN
The efficient scalable syntheses of 2-acetamido-1,2-dideoxy-D-galacto-nojirimycin (DGJNAc) and 2-acetamido-1,2-dideoxy-D-gluco-nojirimycin (DNJNAc) from D-glucuronolactone, as well as of their enantiomers from L-glucuronolactone, are reported. The evaluation of both enantiomers of DNJNAc and DGJNAc, along with their N-alkyl derivatives, as glycosidase inhibitors showed that DGJNAc and its N-alkyl derivatives were all inhibitors of α-GalNAcase but that none of the epimeric DNJNAc derivatives inhibited this enzyme. In contrast, both DGJNAc and DNJNAc, as well as their alkyl derivatives, were potent inhibitors of β-GlcNAcases and β-GalNAcases. Neither of the L-enantiomers showed any significant inhibition of any of the enzymes tested. Correlation of the in vitro inhibition with the cellular data, by using a free oligosaccharide analysis of the lysosomal enzyme inhibition, revealed the following structure-property relationship: hydrophobic side-chains preferentially promoted the intracellular access of iminosugars to those inhibitors with more-hydrophilic side-chain characteristics.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
D-(+)-Glucuronic acid γ-lactone, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad