Merck
Efficacy of adding behavioural treatment or antimuscarinic drug therapy to α-blocker therapy in men with nocturia.
BJU international (2013-03-02)
Theodore M Johnson, Alayne D Markland, Patricia S Goode, Camille P Vaughan, Janet L Colli, Joseph G Ouslander, David T Redden, Gerald McGwin, Kathryn L Burgio
RESUMEN
WHAT'S KNOWN ON THE SUBJECT? AND WHAT DOES THE STUDY ADD?: Nocturia is a common and bothersome lower urinary tract symptom, particularly in men. Many single drug therapies have limited benefit. For men who have persistent nocturia despite alpha-blocker therapy, the addition of behavioural and exercise therapy is statistically superior to anticholinergic therapy. To compare reductions in nocturia resulting from adding either behavioural treatment or antimuscarinic drug therapy to α-adrenergic antagonist (α-blocker) therapy in men. Participants were men who had continuing urinary frequency >8 voids/day) and urgency after 4 weeks of α-blocker therapy run-in and who had ≥1 nightly episode of nocturia. Participants received individually titrated drug therapy (extended-release oxybutynin) or multicomponent behavioural treatment (pelvic floor muscle training, delayed voiding and urge suppression techniques). Seven-day bladder diaries were used to calculate reductions in mean nocturia. A total of 127 men aged 42-88 years with ≥1 nocturia episode per night were included in the study. There were 76 men who had a mean of ≥2 nocturia episodes. Among those with ≥1 nocturia episode, behavioural treatment reduced nightly nocturia by a mean of 0.97 episodes and was significantly more effective than drug therapy (mean reduction = 0.56 episodes; P = 0.01). Participants with ≥2 episodes nocturia at baseline also showed larger changes with behavioural treatment compared with antimuscarinic therapy (mean reduction = 1.26 vs 0.61; P = 0.008). Both behavioural treatment and drug therapy reduced nocturia in men with ≥1 episode of nocturia/night when added to α-blocker therapy. These results were similar even when only those with ≥2 episodes of nocturia were considered. The addition of behavioural treatment was statistically better than bladder-relaxant therapy for nocturia.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Oxybutynin chloride, ≥98% (TLC), powder
Sigma-Aldrich
Oxybutynin chloride, meets EP, USP testing specifications