Merck
The disposition of citric acid cycle intermediates by isolated rat heart mitochondria.
Biochimica et biophysica acta (1981-11-18)
J K Hiltunen, E J Davis
RESUMEN
The mechanism of depletion of tricarboxylic acid cycle intermediates by isolated rat heart mitochondria was studied using hydroxymalonate (an inhibitor of malic enzymes) and mercaptopicolinate (an inhibitor of phosphoenolpyruvate carboxykinase) as tools. Hydroxymalonate inhibited the respiration rate of isolated mitochondria in state 3 by 40% when 2 mM malate was the only external substrate, but no inhibition was found with 2 mM malate plus 0.5 mM pyruvate as substrates. In the presence of bicarbonate, arsenite and ATP propionate was converted to pyruvate and malate at the rates of 14.0 +/- 2.9 and 2.8 +/- 1.8 nmol/mg protein in 5 min, respectively. Under these conditions, 0.1 mM mercaptopicolinate did not affect this conversion, but 2 mM hydroxymalonate inhibited pyruvate formation completely and resulted in an accumulation of malate up to 13.2 +/- 2.9 nmol/mg protein. No accumulation of phosphenolpyruvate was found under any condition tested. It is concluded that malic enzymes but not phosphoenolpyruvate carboxykinase, are involved in conversion of propionate to pyruvate in isolated rat heart mitochondria.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Tartronic acid, ≥97.0%