Merck
Is ATP a substrate for 15-lipoxygenase?
Biochemistry and cell biology = Biochimie et biologie cellulaire (2000-06-30)
R Kumarathasan, F H Leenen
RESUMEN
Lipoxygenases catalyze peroxidation of polyunsaturated fatty acids containing the 1-cis, 4-cis pentadiene structure. Linoleic (18:2), linolenic (18:3), and arachidonic (20:4) acids are the predominant substrates for this class of enzymes. Effects of 15-lipoxygenase on the hydrolysis of adenosine 5'-triphosphate were investigated in vitro using soybean lipoxygenase and adenosine 5'-[gamma-32P]triphosphate. The amount of inorganic phosphate released from adenosine 5'-triphosphate was dependent upon enzyme as well as substrate concentrations, pH, and the duration of incubation. The ATPase activity with a Vmax value of 3.3 mumol.mg protein-1.h-1 and a Km value of 5.9 mM was noted in the presence of different concentrations of ATP at pH = 7.4. Phenidone, a lipoxygenase inhibitor, had no effect on this reaction. These findings suggest that soybean lipoxygenase catalyzes the release of inorganic phosphate from ATP primarily via hydrolysis.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
1-Phenyl-3-pyrazolidinone, 97%