Merck
  • Synthesis and hydrolytic behaviour of glycerol-1,2-diibuprofenate-3-nitrate, a putative pro-drug of ibuprofen and glycerol-1-nitrate.
Synthesis and hydrolytic behaviour of glycerol-1,2-diibuprofenate-3-nitrate, a putative pro-drug of ibuprofen and glycerol-1-nitrate.
The Journal of pharmacy and pharmacology (2001-04-09)
M J Ingram, H A Moynihan, M W Powell, C Rostron
RESUMEN
Nitroxylated derivatives of non-steroidal anti-inflammatory drugs appear to offer protection against the gastrotoxicity normally associated with non-steroidal anti-inflammatory drugs, ostensibly via local production of nitric oxide. A diester of ibuprofen and glycerol-1-mononitrate has been prepared via the condensation of ibuprofen with 3-bromopropan-1,2-diol, followed by silver-(I)-nitrate-mediated nitroxylation. The release of ibuprofen from this diester has been studied in a simulated gastric fluid model with direct analysis by reverse-phase HPLC, using an acetonitrile-water (80%:20%) mobile phase containing trifluoroacetic acid (0.005%). n-Propyl ibuprofen was found to undergo pH-dependent hydrolysis, ranging from negligible hydrolysis at pH 5 to 52% hydrolysis at pH 3, over a 2-h period in this model. The ibuprofen-glycerol mononitrate diester was subjected to the most vigorous model hydrolytic conditions and was found to undergo 50 % hydrolysis during the study period. This study shows that pro-drugs of ibuprofen and glycerol mononitrate can be obtained, and can undergo degradation to the parent drugs under conditions simulating those likely to be encountered in the stomach.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
1-Mononitroglycerin solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®