Merck
Identification and role of ionizing functional groups at the active center of Rhodotorula gracilis D-amino acid oxidase.
FEBS letters (2001-11-07)
L Pollegioni, C M Harris, G Molla, M S Pilone, S Ghisla
RESUMEN
D-Amino acid oxidase (DAAO) is a flavoprotein oxidase that catalyzes the oxidation of amino acids and produces ketoacids and H(2)O(2). The rate of product release from reduced DAAO from Rhodotorula gracilis is pH dependent and reflects a pK(a) of approximately 9.3. Binding of benzoate and 3,3,3-trifluoro-D-alanine to wild-type and Y238F-DAAO is also pH dependent (pK(a)=9.8+/-0.1 and 9.05+/-0.1, respectively for benzoate binding). However, binding of benzoate to Y223F-DAAO is pH independent, indicating the pK(a) is due to Y223-OH. This latter residue is thus involved in substrate binding, and probably is the group that governs product release. In contrast to this, the second active site tyrosine, Y238, has little influence on ligand binding.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
3,3,3-Trifluoro-DL-alanine, 98%