Merck
  • Induction of apoptosis in human blood T cells by 7,8-dihydroneopterin: the difference between healthy controls and patients with systemic lupus erythematosus.
Induction of apoptosis in human blood T cells by 7,8-dihydroneopterin: the difference between healthy controls and patients with systemic lupus erythematosus.
Clinical immunology (Orlando, Fla.) (2003-06-14)
Barbara Wirleitner, Gerlinde Obermoser, Günther Böck, Gabriele Neurauter, Harald Schennach, Norbert Sepp, Dietmar Fuchs
RESUMEN
Neopterin (Neo) and 7,8-dihydroneopterin (H(2)Neo) are produced by human monocyte-derived macrophages upon stimulation with IFN-gamma. Increased amounts of Neo and H(2)Neo in human body fluids are found in many disorders, including viral infections and autoimmune diseases. Recent data suggest that neopterin derivatives may exhibit distinct biochemical functions activating redox-sensitive transcription factors and inducing apoptosis in various cell lines. In this study we investigated the effect of H(2)Neo on human peripheral blood T cells (PBT) from healthy blood donors in comparison with PBT isolated from patients with systemic lupus erythematosus (SLE). H(2)Neo induced apoptosis in healthy PBT in a concentration-dependent manner. In short time culture, a significantly lower ability of PBT isolated from patients with SLE to undergo apoptosis in response to H(2)Neo compared to healthy controls was detected. Our results suggest a possible role of the neopterin derivative H(2)Neo in T cell apoptosis mediated by stimulated monocytes/macrophages.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
7,8-Dihydroneopterin, ≥97.0% (HPLC)