Merck
  • Differences in the in vivo dynamics of neurotransmitter release and serotonin uptake after acute para-methoxyamphetamine and 3,4-methylenedioxymethamphetamine revealed by chronoamperometry.
Differences in the in vivo dynamics of neurotransmitter release and serotonin uptake after acute para-methoxyamphetamine and 3,4-methylenedioxymethamphetamine revealed by chronoamperometry.
Neurochemistry international (2005-06-28)
Paul D Callaghan, Rodney J Irvine, Lynette C Daws
RESUMEN
Illicit use of p-methoxyamphetamine (PMA) is rapidly increasing. However, little is known about the acute effects of PMA on neurotransmission in vivo. High-speed chronoamperometry was used to monitor neurotransmitter release and clearance in anesthetized rats after local application of PMA or 3,4-methylenedioxymethamphetamine (MDMA). In striatum, PMA caused less neurotransmitter release than MDMA. PMA-evoked release could be partially blocked by pre-treatment with a serotonin (5-HT) reuptake inhibitor, suggesting that evoked 5-HT release contributed to the electrochemical signal and was mediated by the 5-HT transporter (SERT). MDMA-evoked release was not blocked by a SERT inhibitor, suggesting that primarily DA was released. To study the effect of these amphetamines on clearance of 5-HT mediated specifically by the SERT, clearance of exogenously applied 5-HT was measured in the CA3 region of the hippocampus. In contrast to the striatum where 5-HT is cleared by both the SERT and the dopamine transporter (DAT), 5-HT is cleared primarily by the SERT in the CA3 region. This is also a region where neither PMA nor MDMA evoked release of neurotransmitter. The maximal inhibition of 5-HT clearance was greater after PMA than MDMA. These data demonstrate in vivo (1) brain region variability in the ability of PMA and MDMA to evoke release of neurotransmitter; (2) that clearance of 5-HT in the striatum is mediated by both the SERT and the DAT; (3) distinct differences in the amount and nature of neurotransmitter released in the striatum after local application of PMA and MDMA and (4) that PMA is a more efficacious inhibitor of 5-HT clearance in the hippocampus than MDMA. These fundamental differences may account for the more severe adverse reactions seen clinically after PMA, compared to MDMA.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
PMA hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®